NANOCOLOR® Photometric Tube Tests, Total Chromium
Supplier: CTL SCIENTIFIC SUPPLY CORP
985059
77578-588EA 130.81 USD
77578-588
NANOCOLOR® Photometric Tube Tests, Total Chromium
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of chromium in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.