NANOCOLOR® Photometric Tube Tests, pH
Supplier: CTL SCIENTIFIC SUPPLY CORP
91872
77578-552EA 64.07 USD
77578-552
NANOCOLOR® Photometric Tube Tests, pH
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of pH in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.