NANOCOLOR® Photometric Tube Tests, Fluoride
Supplier: CTL SCIENTIFIC SUPPLY CORP
985040
77578-496EA
63.25
USD
77578-496
NANOCOLOR® Photometric Tube Tests, Fluoride
Environmental Test Kits
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of fluoride in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in ext.Ø 16 mm tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)