NANOCOLOR® Photometric Tube Tests, Chromate
Supplier: CTL SCIENTIFIC SUPPLY CORP
985024
77578-452EA
87.86
USD
77578-452
NANOCOLOR® Photometric Tube Tests, Chromate
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of chromate in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in ext.Ø 16 mm tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)