Slide Drying Hotplate, Slimline
Supplier: StatLab
SI-SDH-SLIM
77578-310EA 1655.37 USD
77578-310
Slide Drying Hotplate, Slimline
Slide Warmers/Dryers
The StatLab slimline slide drying hotplate is designed to efficiently dry slides through the use of heated air radiating from a hotplate beneath the slides.
  • Variable digital temperature control
  • Ultra-fast heating system for rapid warm up
  • Movable rails
  • Anti-microbial powder coat finish
  • Memory function for desired temperature at start-up, integrated circuit breakers
An ultra-fast heating system ensures rapid warm up, and digital controls provide temperature accuracy. Microscope slides are supported on four movable rails which allow heated air to pass beneath the slides to accellerate drying, while slides sit at a sloped angle to encourage water drain off.
The rails can be removed from the instrument so that slides can be placed directly on the heated surface when required.
Ordering information: An optional stainless steel lid (sold separately) is available which can be used to further reduce drying times and efficiency.