Paraffin Dispenser, Slimline
Supplier: StatLab
SI-PWD-SM
77578-304EA 2942.87 USD
77578-304
Paraffin Dispenser, Slimline
Paraffin Wax Trimmers and Dispensers
The novel design of the slimline paraffin dispenser ensures a minimal requirement for bench space, while maintaining a large 7.5 L capacity tank for melting up to 6 kg of pelletized paraffin at a time.
  • High tap position (153 mm from the bench) allows larger sized containers to be used, integral tap filter screen, heated tap
A digital temperature control provides a highly accurate temperature setting, and is coupled with an ultra-fast heating system for rapid melting of pelletized paraffin, available for dispensing after only 30 minutes.