X-Ray and Metal Detectable Micro Spatulas
Supplier: SAMPLING SYSTEMS LTD
M1013B-170
77578-202EA 9.88 USD
77578-202
X-Ray and Metal Detectable Micro Spatulas
Spatulas
The X-ray and metal detectable micro spatulas are ideal for use in food and pharmaceutical production and quality control work. The spatulas are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the powder in line with HACCP guidelines.
  • Robust PP construction
  • PP gives excellent chemical resistance to many products
  • Long ergonomic handle
  • Full batch traceability
The spatula are ideal for sampling from bags and surface sampling from containers aswell as general work. The narrow blade makes them ideal for use with small amounts of powder.
To ensure the cleanliness of the sampling spatula they are manufactured and packed in a cleanroom. They can be used as a single use disposable product straight from the bag or reused.
Certifications: These spatula conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Packaging: Supplied individually bagged.