Micro Spatula
Supplier: SAMPLING SYSTEMS LTD
A106-20
77577-632EA 55.18 USD
77577-632
Micro Spatula
Spatulas
Designed for measuring and sampling small amounts of powder.
  • Ideal for surface sampling and dispensing small quantities
  • Crevice-free and easily cleaned
  • Available from stock
  • Fast delivery
High-quality micro spatulas are excellent for a variety of applications, such as measuring, sampling, or dispensing small quantities of powder, granule, and cream. They are crevice-free and therefore easily cleaned.