SteriWare® Powder Funnels
Supplier: SAMPLING SYSTEMS LTD
8111H-50 8111H-50S 8111P-350 8111P-350S 8111H-100 8111H-100S 8111H-200 8111H-200S
77577-066EA 53.52 USD
77577-066 77577-068 77577-070 77577-072 77577-058 77577-060 77577-062 77577-064
SteriWare® Powder Funnels
Funnels Powder Funnels
These single use powder funnels made from chemically resistant, food grade HDPE are ideal for use with powders, creams and liquids.
  • Available as sterile and non sterile versions
  • Manufactured under cleanroom conditions: Can be used straight from the bag
  • Cross-contamination risk is eliminated
  • No cleaning is required before or after sampling
The wide opening at the base of the funnel will help prevent powder blockages.
Certifications: Material conforms to FDC and EC standards. Full batch traceability.
Packaging: Individually bagged.