PureBag Sample Bags
Supplier: SAMPLING SYSTEMS LTD
8080A-450
77576-998EA 389.1 USD
77576-998
PureBag Sample Bags
Bags Sample Bags
These are an ideal low cost alternative sterile bags for standard containers and are available in wide range of sizes.
  • Pyrogen free
  • Suitable up to 80 °C
  • Made from a special tough grade of LDPE
  • Leakproof with unique metal closure system
Certifications: Complies with FDA, USDA and HACCP.