Tablet and Capsule Sampler
Supplier: SAMPLING SYSTEMS LTD
4010B-01
77576-388EA 3359.56 USD
77576-388
Tablet and Capsule Sampler
Samplers Solid Samplers
Our tablet sampler is design to take a cross sectional sample of tablets and capsules in a container. Its is designed to take samples at 5 points through the container. A handy handle makes it easy to manoeuvre and operate the sampler.
  • Made from quality 316L stainless steel
  • The sampler body is then electropolished afterwards to give a clean bright polish finish
  • As standard it is available in 1 m length
Put the sampler into the container before its is filled. When filled the sampler will automatically take samples from various levels throughout the container. After removing the sampler the end can be lowered to release the capsule and tablets.