Cohesive Sampler
Supplier: SAMPLING SYSTEMS LTD
2000S-01
77576-356EA 1134.4 USD
77576-356
Cohesive Sampler
Samplers Solid Samplers
This cohesive sampler is designed for powders.
  • For taking single point samples
  • Handle and four sample cells
  • 0,25, 0,50, 0,75, 1,0 ml unit doses
Used throughout the pharmaceutical industry from sticky (or cohesive) powders.
Delivery information: A nylon bristle can be ordered separately.
Packaging: In a case.