U-D Master Sleeve Samplers
Supplier: SAMPLING SYSTEMS LTD
1500A-01 1500W-17 1500W-18 1500W-22 1500A-02 1500A-03 1500A-04 1500S-01 1500W-01 1500W-02 1500W-03 1500W-04 1500W-08 1500W-12
77576-264EA 209.43 USD
77576-264 77576-292 77576-294 77576-296 77576-266 77576-268 77576-270 77576-272 77576-274 77576-276 77576-278 77576-280 77576-282 77576-284
U-D Master Sleeve Samplers
Samplers Solid Samplers
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
  • Quick and easy accurate unit dose sampling
  • Different volumes can be taken using the same sampler
  • Simply change the tip for a different volume
Designed for use with free flowing powders and granules.