Sack Samplers
Supplier: SAMPLING SYSTEMS LTD
1320A-01 1320B-01 1320C-01
77576-232EA 155.2 USD
77576-232 77576-234 77576-236
Sack Samplers
Samplers Solid Samplers
These sack samplers are designed to take fast powder samples through the sides of sacks and bags.
  • Suitable for sampling free flowing powders and granules
  • 316L stainless steel construction
  • Unique range of sample collection methods available
Insert the sampler through the side of the sack. Angle sampler downwards to allow the product to flow into the sample collecting container and seal the hole using an ultra adhesive label.
Ordering information: Includes a 250 ml sampling bottle.