Powder Samplers
Supplier: SAMPLING SYSTEMS LTD
1220J-2000 1220J-900 1220A-1500 1220A-600 1220A-900 1220B-1500 1220B-300 1220B-600 1220B-900 1220J-1500
77576-086EA 2793.47 USD
77576-086 77576-088 77576-070 77576-072 77576-074 77576-076 77576-078 77576-080 77576-082 77576-084
Powder Samplers
Samplers Solid Samplers
Pocket samplers for discrete zone samples from powders and granules.
  • Simple operation and robust design
  • Stainless steel or PTFE inner rod
  • Can be stripped and cleaned very quickly
Suitable for use in pharmacy applications.
Delivery information: Supplied with pointed tips as standard.