Powder Max Manual Sampler
Supplier: SAMPLING SYSTEMS LTD
1213A-01
77576-062EA 636.07 USD
77576-062
Powder Max Manual Sampler
Samplers Solid Samplers
The Powder Max is ideal for taking large samples of free-flowing powders.
  • Takes large samples
  • Can be fully dismantled for easy cleaning
  • Easy operation
This single point sampler is ideal for sampling large volumes of free-flowing powders and granules. The intuitive design and self closing sleeve means the sampler remains closed until it is pulled out. After use, the sampler can be fully dismantled for easy cleaning, in line with GMP guidelines.