Spatula, V Shape
Supplier: SAMPLING SYSTEMS LTD
1095S-150
77576-040EA 121.43 USD
77576-040
Spatula, V Shape
Spatulas
This V spatula made from stainless steel 316 is ideal for surface sampling of powders and core sampling of viscous material like cheese, pastes and gels.
  • Polished finish
  • Fully welded, crevice free construction
  • ‘V’ shape allows easy discharge of the powders into containers or bags