PowderThief Manual Samplers
Supplier: SAMPLING SYSTEMS LTD
1030A-2000 1030AT-200 1030B-2000 1030CT-20
77575-950EA 624.55 USD
77575-950 77575-966 77575-972 77575-994
PowderThief Manual Samplers
Samplers Solid Samplers
Widely used in pharma, the PowderThief is ideal for sampling free-flowing powders.
  • World leader in powder sampling
  • Suitable for sampling free-flowing powders
  • Range of tips available
  • Made from 316L stainless steel
The longstanding PowderThief takes samples of free-flowing powders and granules in a wide range of sample volumes. The sampler is easy to use and comes in a wide range of lengths. Simply push the sampler into the powder and push down on the handle to take the sample. Move the body downwards to then close in the sample before removing and emptying the tip.