Oberon 12 Cal TCG™ Arc Flash Face Shield and Hard Cap
Supplier: Sellstrom
21AGR12AF-C+500
77575-524EA 182.26 USD
77575-524
Oberon 12 Cal TCG™ Arc Flash Face Shield and Hard Cap
Face Shields
Shield made from a durable polycarbonate and has anti-fog and scratch-resistant coatings.
Face shield comes equipped with an Oberon or Canadian hard cap.
Certifications: The Oberon 12 Cal Arc Flash Face Shield meets ANSI /ISEA 125 Level 2 Conformity and Arc Flash PPE Category 2 Standards with an arc rating of 17 cal/cm2. Please refer to NFPA 70E or CSA Z462 Standards for specific selection requirements.