Corning® Stripettor® Pro Pipette Controller
Supplier: Corning
4999
77575-370EA 469.5 USD
77575-370
Corning® Stripettor® Pro Pipette Controller
Pipettes Pipette Controllers
This light, ergonomic, motorized pipet controller is designed for use with glass and plastic serological pipets from 0.5 to 100 ml.
  • Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities
  • Aspirating and dispensing speed is controlled through the finger triggers
  • When fully charged, the battery enables up to 6 hours of continuous use
  • LCD display for continuous visualization of speed, modes, and battery status
  • The speed can be set to one of 3 values for both HIGH and LOW value
Delivery information: Supplied with a universal power supply, 0.2 μm PTFE filter, a wall hanger and a charging stand.