Monarch® Spin Columns S2D and Tubes
Supplier: New England Biolabs (NEB)
T1117L
77575-360EA 94 USD
77575-360
Monarch® Spin Columns S2D and Tubes
Filters Centrifugal Filters
Monarch® Spin Columns S2D and Tubes have been designed and assembled to deliver high performance for nucleic acid purification and concentration.
  • Includes 100 spin columns and 100 collection tubes designed for nucleic acid purification
  • Minimal to no buffer retention/risk of carry-over contamination
  • Low elution volume (≥30 µl) from unique column design
  • 20 to 100 µg nucleic acid binding capacity, depending on application
  • Made with less plastic than conventional columns from leading suppliers
Monarch® spin columns and tubes are available for purchase separately and are included with Monarch® Spin Nucleic Acid Purification Kits. The columns are designed without an O-ring, eliminating buffer retention and the risk of salt carry-over contamination, resulting in highly pure nucleic acid. The tapered column design allows for elution volumes as low as 30 µl for highly concentrated nucleic acid suitable for various downstream applications. Additionally, these spin columns are designed with sustainability in mind, using less plastic than conventional columns without compromising performance. These columns are compatible for centrifugation and vacuum manifold protocols and are supplied with collection tubes.