DNase I-XT Reaction Buffer
Supplier: New England Biolabs (NEB)
B0570S
77575-352EA 27 USD
77575-352
DNase I-XT Reaction Buffer
Buffers Cloning Buffers
New England Biolabs supplies a 10X reaction buffer with all of its enzymes.
  • For use with DNase I-XT (M0570)
  • Supplied in easy to use 10X format
At a 10X concentration this reaction buffer assures optimal activity of the enzyme.