Moticam BMH4000X Digital Camera HDMI
Supplier: Motic
1100600101731
77575-322EA 1739.67 USD
77575-322
Moticam BMH4000X Digital Camera HDMI
Cameras
The Moticam BMH4000X is a C-mount multitasking microscopy camera, that delivers an impressive 1080p full HD live image with maximum color fidelity on an integrated LCD screen. It does not require a computer for standard operation, as it comes with an on-board software that will allow you to use it through its 11.6" full HD integrated screen, with a mouse connected to the USB port of the camera.
  • Impressive 1080p full HD live image
  • Video output level: 4K (30 fps @ 3840×2160) H264/H265 encoded MP4 file in SD card
Unlike other cameras, the Moticam BMH4000X can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through the built-in 11.6" LCD screen, or connect your camera through USB to a Windows, Mac or Linux computer.
By having an integrated SD card slot, not only does saving images not require a computer, but it also allows flexibility to save portable data.
Delivery information: Package includes 12 V power adapter, CS ring, Motic four-dot calibration slide, HDMI cable, manual, 12 mm lens, adapter φ30/φ38, macro tube.