Moticam 4000X Digital Camera HDMI
Supplier: Motic
1100600101641
77575-320EA 1333.85 USD
77575-320
Moticam 4000X Digital Camera HDMI
Cameras
The Moticam 4000X is dedicated to documentation of microscopic results with a clear focus on maximum color fidelity and fast live image. It does not require a computer for standard operation, as it comes with an on-board software that will allow you to use it through its HDMI port, and with a mouse connected to the USB port of the camera.
  • The HD affordable multi-tasking microscopy camera
Unlike other cameras, the Moticam 4000X can be connected to a computer or through HDMI. Simply plug the camera through the HDMI port and display images with the use of the onboard software and a USB mouse through an HDMI monitor, or connect your camera through USB to a Windows, Mac or Linux computer.
By having an integrated SD card slot, not only does saving images not require a computer, but it also allows flexibility to save portable data.
Ordering information: Package includes 12 V power adapter, CS ring, Motic four-dot calibration slide, HDMI cable, manual, 12 mm lens, adapter φ30/φ38, macro tube.