DepleteX® Mitochondrial DNA Depletion Kit
Supplier: REVVITY HEALTH SCIENCES, INC.
NOVA-512890
77575-228EA 1236 USD
77575-228
DepleteX® Mitochondrial DNA Depletion Kit
Nucleic Acid Reagents Next Generation Sequencing Reagents
The DepleteX® mitochondrial DNA depletion kit employs a straightforward CRISPR-based method to selectively remove unwanted human molecules derived from mitochondrial nucleic acid before sequencing.
  • Efficiently remove mitochondrial reads from DNA or RNA samples
  • Kit enhances sequencing by eliminating mitochondrial reads from DNA or RNA NGS libraries before sequencing
Efficiently remove mitochondrial reads from DNA or RNA samples. Mitochondrial reads consume significant sequencing space in certain sample type and applications. Depleting these reads prior to sequencing allows redirection of sequencing resources toward valuable information for researchers. The DepleteX® Mitochondrial DNA depletion kit enhances sequencing by eliminating mitochondrial reads from DNA or RNA NGS libraries before sequencing.
The DepleteX® mitochondrial DNA depletion kit is used after DNA or RNA library preps and prior to sequencing. This kit is compatible with most DNA-seq and RNA-seq workflows; however, it is recommended using it with the NEXTFLEX™ Rapid XP V2 DNA-seq kit or NEXTFLEX™ RNA-seq 2.0 kit.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.