SHOWA 383 Biodegradable Multi-Purpose Gloves
Supplier: Showa
383L-08 383M-07 383S-06 383XL-09 383XXL-10
77575-114CS 307.93 USD
77575-114 77575-142 77575-108 77575-156 77575-116
SHOWA 383 Biodegradable Multi-Purpose Gloves
Gloves
The SHOWA 383 is the world’s-first biodegradable multi-purpose glove! Its reduced thickness ensures the 383 optimum tactility when working on delicate projects. In addition, the design provides comfort and flexibility during extended wear. The 383 is manufactured with the Eco Best Technology®, making it biodegradable.
  • General purpose
  • Thin palm material
  • Thin finger thickness
  • Embossed microporous nitrile coating
  • Lightweight and flexible
  • Engineered yarn
  • Reusable and Biodegradable
The gloves are abrasion and oil resistant, latex free with increased dexterity and sensitivity. It has an optimal liner design.
Trades and Applications: Intricate parts handling, light assembly of oil-coated pieces, mechanical and engineering, automotive repairs and maintenance, shuttering, component assembly, heavy equipment operation, tool operation and transporting.
Industries: Construction, mechanical, aerospace, automotive, engineering, mining, airports and ports.
Certifications: CE, EN 388:2016+A1:2018.
Packaging: 12 Pair per polybag, 12 Polybags per case, 144 Pair per case.