SHOWA Duracoil 386 Cut-Resistant Gloves
Supplier: Showa
386L-08 386M-07 386S-06 386XL-09 386XXL-10
77575-180CS 623.52 USD
77575-180 77575-112 77575-182 77575-096 77575-130
SHOWA Duracoil 386 Cut-Resistant Gloves
Gloves
The DURACoil 386 utilizes advanced HPPE reinforced lining to provide cut and abrasion resistance, without sacrificing dexterity. The microporous coating disperses oil to increase grip. So even for tasks on a smaller scale, such as component assembly, the 386 enables a full range of movement and control.
  • Ergonomic and optimal liner design with embossed microporous nitrile coating
  • Form-fitting
  • Oil-resistant
  • Extra grip and flexible
  • Increased dexterity
  • Breathable
  • Seamless knit
They are machine washable.
Trades and applications: Intricate parts handling, brick laying, flashings and seals, maintenance, steel fixing, component assembly.
Industries: Aerospace, automotive, engineering, manufacturing and mechanical.
Certifications: CE, EN388:2016+A1:2018, ANSI/ISEA 105-2016 CUT.