Cut Resistant Gloves, XC800
Supplier: Showa
XC800L-08 XC800M-07 XC800S-06 XC800XL-09 XC800XXL-10
77575-166CS 528.34 USD
77575-166 77575-152 77575-090 77575-094 77575-168
Cut Resistant Gloves, XC800
Gloves
The new ultra-light XC800 combines high cut-resistance level E/A5 provided by a seamless HPPE/stainless steel/pester liner with a durable, oil-resistant microporous nitrile palm coating usable in various industrial applications.
  • HPPE reinforced
  • Ergonomic
  • Microporous nitrile coating
  • Cut-resistant, abrasion-resistant
  • Flexible, breathable
  • Lightweight, form-fitting
  • Increased dexterity
  • Seamless knit
Application: Light assembly of oil-coated pieces, maintenance, mechanical and engineering, repair glass and windows, handling heavy equipment, intricate parts handling.
Certifications: ANSI/ISEA 105-2016 CUT, EN 388:2016.
Packaging: Pair per polybag: 12; Polybags per case: 6; Pair per case: 72.