Genevac MV System Connection Kit for MV Centrifugal Concentrator
Supplier: Genevac
100409291
77575-000EA 589.86 USD
77575-000
Genevac MV System Connection Kit for MV Centrifugal Concentrator
Concentrators
miVac centrifugal evaporation systems provide a compact, high performing solution for removing water and organic solvents from a variety of sample formats. Configure a system to meet your application requirements.
  • Concentrators control the pump, heat and run time with a simple user interface and large digital display
  • Quiet electro-magnetic drive ensures maintenance free operation
  • Choose from two centrifuge bowl sizes, three pumps and add-on accessories
  • Variety of sample holder rotors, ordered separately accommodate tubes, micro tubes, vials or microplates
System connection kits have everything you need for connection of miVac pumps and/or cold traps to your concentrator. Includes, 1 and 1.5 m lengths ½" wire reinforced tube, power lead to control pump and catch pot for pump. Extension kit is also available.