Genevac MV MST Defrost Free Condenser for Centrifugal Concentrator
Supplier: Genevac
100403917
77574-990EA 5602.13 USD
77574-990
Genevac MV MST Defrost Free Condenser for Centrifugal Concentrator
Concentrators
MV MST-12060 SpeedTrap condenser. Cold traps condense solvent vapours back to liquid with a corresponding massive volume reduction. This volume reduction can greatly improve the performance of any vacuum system by helping to maintain a vacuum and speeding up the concentration process considerably.
  • Compact refrigerated condenser, makes efficient use of bench space
  • Uses a greener refrigerant, R1270
  • Defrost free trap with internal gas compressor and 1 L glass collection jar
  • Three defrost modes to select from depending on the solvent
Designed for ultimate performance, the MV SpeedTrap cold condenser coils are suspended in the vapor path so solvent vapours condense directly onto the coils and ultimately run off into the collection vessel below for a compact, efficient unit offering excellent visibility and ease of use.
Choose from three settings to suit your application needs: Auto defrost - for collection of solvents liable to freeze the system periodically defrosts preventing build-up of ice on the coils, Continuous chilling - for maximum recovery of solvents that do not freeze above −50 °C including most organic solvents, and Manual defrost - when continuous chilling is used with solvents above −50 °C and ice accumulates on the coils or when freeze drying samples.
The MV SpeedTrap can be used in conjunction with a Super Vacuum pump to freeze dry up to 250 ml of water. Pre- frozen samples can be placed in the cascade condenser jar provided with the freeze drying accessory kit.