Roto-Rock Genie Rotator Rocker, 120 V

Supplier: Scientific Industries
SI-R100
77574-960EA 2152.5 USD
77574-960
Roto-Rock Genie Rotator Rocker, 120 V
Shakers and Mixers Rotating/Rocking Shakers
Programmable combination rotator rocker with digital touch screen, featuring magnetic platform and clip plate system.

  • 120 V model
  • Intuitive touch screen control makes changing program settings easy
  • Setting the tilt angle and speed control provides thorough mixing while eliminating spills when using open containers in rocking mode
  • The two-sided magnetic platform holds virtually any type of tube and other sample vessels used in the laboratory when using the GENIE clip plates (sold separately)
  • Compact design saves valuable bench space in the laboratory
  • Designed for use in cold rooms, incubators (non-CO₂), or at ambient temperature
  • 2 year warranty

Programable all-in-one benchtop GENIE Rocker/Rotator or cycle from Rock to Rotate. Digital set/control for rpm, total time, cycle time. Features 'pause/resume' button allowing the operator to add reagents to a sample during processing.

Programming modes: Rock, rotate, or timed auto-switching between rock and rotate
Rocking tray size: 300×300 mm
Power: DC 24 V, 2.5 A

Certifications: CE certified.

Delivery information: Supplied with magnetic platform and non-slip rocking tray. Accessories must be ordered separately: GENIE clip plate systems designed to handle a variety of vessels from PCR tube, vials, bottles and sealed bags.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR