PermaShield™ Matte Polyester Label for BradyJet™ J2000 Printers
Supplier: Brady Worldwide
175617
77574-948RL 544.59 USD
77574-948
PermaShield™ Matte Polyester Label for BradyJet™ J2000 Printers
Labels
Upgrade the durability and longevity of your identification projects, even in the toughest environments, with PermaShield™ polyester labels designed for BradyJet printers.
  • Simply print label, adhere to surface and apply the matching overlaminate
  • Commonly used to print product labeling, logos, right to know labels and barcode applications
  • Works best on flat, smooth surfaces, indoor use
  • An ink cartridge is required to print this material; use part number J20-CMY
  • Abrasion-resistant, chemical-resistant, fade-resistant, oil-resistant, solvent-resistant
The durable polyester label material (B-2475) plus polyester overlaminate (B-996) protects against numerous harsh solvents and provides a more glossy appearance.
Delivery information: PermaShield by Brady is a 2-in-1 package that includes a printable label plus matching clear polyester overlaminate.