SafeCup™ III 11-Panel All-in-One Cup Tests
Supplier: Germaine Laboratories
41011
77574-668CS 200.5 USD
77574-668
SafeCup™ III 11-Panel All-in-One Cup Tests
Drug Tests
Rapid qualitative tests for the detection of drugs of abuse in urine.
  • A total collection, testing, and transport system
  • Easier to use, easier to read
  • Quick results in 5 minutes
  • Includes 'Click Seal' cap for improved leak resistance
  • Store at room temperature
  • Adheres to SAMHSA cutoff levels
  • Includes gloves, temperature strip, security seals, IFU
All-in-One cup tests for amphetamine, barbiturates, benzodiazpines, buprenorphine, cocaine, methadone, mdma, methamphetamine, morphine, oxycodone, and phencyclidine. 25 cups per case. CLIA waived.
Certifications: CLIA waived for over the counter and point of care use.
Ordering information: Orders received by 12:30 PM CST process same day. If not in stock, product requires 1 to 2 days lead time.