Logic® Particle Counter Integration Kit
Supplier: Labconco
3034000
77574-624EA 255 USD
77574-624
Logic® Particle Counter Integration Kit
Particle Counters
Particle counter integration kit for Logic Biosafety cabinet. Compatible with 4 to 20 mA output.
  • For use with user supplied particle counter
  • Applicable to Logic Biosafety cabinets 2024 or newer models
  • 115 V, 15 A
  • Weight: 3.0 lbs.
  • Dimensions: 2W×29D×1" H