Logic® Mag-Zone™ Waste Bag Holder
Supplier: Labconco
3033100
77574-620EA 204 USD
77574-620
Logic® Mag-Zone™ Waste Bag Holder
Racks Bag Racks
Logic Mag-Zone waste bag holder holds one standard benchtop biowaste bag.
The Mag-Zone magnetic waste bag holder provides easy and safe receptacle area to hold one standard benchtop biowaste bag. It is stainless steel in design and is for use inside the Labconco Logic biosafety cabinet.