Logic® Mag-Zone™ Magnetic Pipette Rack
Supplier: Labconco
3033000
77574-618EA 255 USD
77574-618
Logic® Mag-Zone™ Magnetic Pipette Rack
Pipette Stands
Logic Mag-Zone magnetic pipette rack holds four standard lab pipettes and one standard pipette aid.
The Mag-Zone magnetic pipette rack allows for organization capabilities inside your cabinet. It holds four standard lab pipettes and one standard pipette aid. Engineered from stainless steel it can easily be decontaminated. This magnetic accessory is designed for use inside the Labconco Logic biosafety cabinet.