Airflow Sensor for Recirculating Logic Class II Type A2 Biosafety Cabinet
Supplier: Labconco
3277600
77574-596EA 510 USD
77574-596
Airflow Sensor for Recirculating Logic Class II Type A2 Biosafety Cabinet
Gas Detection Systems
Airflow sensor for Logic Class II Type A2 Biosafety cabinet. Requires installation and certification.
  • Display inflow/downflow on Logic screen
Logic A2 Airflow Sensor allows display of inflow and downflow velocities on screen.