UV Lamp and Ballast
Supplier: Labconco
3289100
77574-552EA 340 USD
77574-552
UV Lamp and Ballast
Lights UV Lights, Lamps and Bulbs
UV lamp and ballast for L4 Logic biosafety cabinets.
  • 254 nm UV lamp and ballast
  • Compatible with all voltages
  • Included in Accessory Package
  • Shipping weight 2.3 kg (5 lbs.)
The UV Lamp kit is designed for the field installation of a UV lamp into cabinets that were not factory equipped for UV. This is engineered for the 2024 Logic Biosafety Cabinets and PuriCare procedure stations. Installation is required.