External Thermal Printer for Vertical Autoclaves
Supplier: TUTTNAUER USA CO. LTD.
THE002-0116
77574-048EA 869.2 USD
77574-048
External Thermal Printer for Vertical Autoclaves
Printers Document Printers
External Thermal printer for Vertical LabSci autoclaves. Printer is capable of printing barcode labels or sterilizer cycle information.
Prints cycle data on thermal paper or UDI barcode labels.
Ordering information: The thermal paper (77574-052) is not included and must be ordered separately. Supplied with 1 roll of labels (77574-050).