VEE GEE Water Testing Field Kit
Supplier: Heathrow Scientific
43036-KIT
77574-036EA 158.84 USD
77574-036
VEE GEE Water Testing Field Kit
Refractometers Handheld Refractometers
Convenient and portable.
  • Testing where and when you need it
  • ATC
This water testing field kit includes all the necessary components for convenient on-site testing of water salinity measurements wherever and whenever needed.
Delivery information: Each kit contains a handheld refractometer, sample cup, transfer pipette, durable carrying case, microfiber cloth, bottle for distilled water, and calibration screwdriver.