VEE GEE Honey Moisture Refractometer Field Kit
Supplier: Heathrow Scientific
43006-KIT
77574-034EA 158.84 USD
77574-034
VEE GEE Honey Moisture Refractometer Field Kit
Refractometers Handheld Refractometers
Convenient and portable.
  • Take honey, water and baume measurements right at the hive
  • ATC
This refractometer field kit includes all the necessary components for convenient on-site testing of honey measurements wherever and whenever needed.
Delivery information: Each kit contains a handheld refractometer, sample cup, transfer pipette, durable carrying case, microfiber cloth, bottle for distilled water, and calibration screwdriver.