One-Minute Competent Cells (DH5 Alpha)
Supplier: ReVacc Scientific
L2A1-8 L2A1-96
77574-024EA 56.1 USD
77574-024 77574-026
One-Minute Competent Cells (DH5 Alpha)
Competent Cells
The one-minute competent cells (DH5 Alpha) are engineered for chemical transformation with high efficiency.
  • One-Minute competent cells are highly efficient (>10E8 transformants/µg pUC19)
  • No need for heat shocking or followed shaking as traditional transformation
  • One-minute processing time and directly spread on solid medium (agar plate)
  • Broad utility for general cloning, blue-white screening, or plasmid isolation
  • Multiple formats for high-throughput applications
Just mix DNA with this competent cell for 1 minute and spread on plate. No heat shock, no shaking needed. This product can be used for cloning, sub-cloning, synthesized DNA, or PCR fragment assembly, library construction and more. 50 µl per PCR tube.
Packaging: Shipped with dry ice for overnight delivery. Stored at –80 °C.
Caution: This product is intended for research use only.