n-Dotriacontane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dotriacontane
A822765-1G
A822765-25G
A822765-5G
A822765-100G
A822765-500G
76807-612EA
14
USD
76807-612
76807-614
76807-616
77573-618
77573-620
n-Dotriacontane 98%
n-Dotriacontane
Caution: for Research Use Only
Formula:
H₃C(CH₂)₃₀CH₃ |
MDL Number:
MFCD00009411 CAS Number: 544-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...