1,6-Hexanedithiol 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,6-Dimercaptohexane
A808980-5G
A808980-25G
77199-678EA
17
USD
77199-678
77573-584
1,6-Hexanedithiol 98%
1,6-Hexanedithiol
Caution: for Research Use Only
Formula:
C₆H₁₄S₂ MW: 150.31 g/mol Boiling Pt: 242 Melting Pt: -21 |
MDL Number:
MFCD00004910 CAS Number: 1191-43-1 UN: 3334 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...