Benzophenone 98%

Supplier: Ambeed
Danger

Synonyms: Diphenylmethanone, Diphenyl ketone

A704460-25G A704460-500G A704460-1000G A704460-100G
77158-472EA 5.15 USD
77158-472 77158-474 77552-138 77573-286 77573-288
Benzophenone 98%
Benzophenone

Caution: for Research Use Only

Formula: C₁₃H₁₀O
MDL Number: MFCD00003076
CAS Number: 119-61-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR