2,3,5-Trifluoropyridine 98%

Supplier: Ambeed
Danger

A667946-1G A667946-5G A667946-25G
103840-484EA 27 USD
103840-484 76844-688 76844-690 77573-154
2,3,5-Trifluoropyridine 98%
2,3,5-Trifluoropyridine

Caution: for Research Use Only

Formula: C₅H₂F₃N
Boiling Pt: 102 °C (760 mm Hg)
Flash Pt: 30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001162
CAS Number: 76469-41-5
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR