4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 98%

Supplier: Ambeed
Warning

A352479-250MG A352479-100MG
77568-710EA 29.87 USD
77568-710 77572-036 77626-680
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) 98%
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)

Caution: For research use only.

Formula: C₁₆H₁₅N₅·2HCl
MDL Number: MFCD00012681
CAS Number: 28718-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR