2-Piperazinone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Piperazin-2-one
A319699-25G
A319699-5G
A319699-100G
A319699-10G
A319699-500G
A319699-1G
103836-146EA
6
USD
103836-146
76838-996
76838-998
77114-708
77114-710
77114-712
77571-908
2-Piperazinone 98%
2-Piperazinone
Caution: for Research Use Only
Formula:
C₄H₈N₂O MW: 100.12 g/mol Boiling Pt: 307.1 °C (760 mm Hg) Melting Pt: 136…140 °C Flash Pt: 166.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318687 CAS Number: 5625-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...