4-(2-Pyridyl)benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde

A274897-10G
103813-232EA 5 USD
103813-232 103853-160 77571-688
4-(2-Pyridyl)benzaldehyde 98%
4-(2-Pyridyl)benzaldehyde

Caution: Keep in a dark place, sealed in dry, room temperature

Formula: C₁₂H₉NO
Melting Pt: 50…52 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD01863537
CAS Number: 127406-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR