(R)-tert-Butyl-2-aminopropanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A124410-10G
A124410-25G
A124410-1G
A124410-100G
103800-526EA
6
USD
103800-526
103831-368
77108-752
77108-754
77312-662
77571-466
(R)-tert-Butyl-2-aminopropanoate hydrochloride 97%
(R)-tert-Butyl-2-aminopropanoate hydrochloride
Caution: for Research Use Only
Formula:
C₇H₁₅NO₂·HCl Melting Pt: 170…175 °C Flash Pt: 78.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077087 CAS Number: 59531-86-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...